BugBuster® 10X Protein Extraction Reagent, MilliporeSigma
Supplier: MilliporeSigma
70921-4 70921-5
CA80508-846EA 1044.28 CAD
CA80508-846 CA80509-636
BugBuster® 10X Protein Extraction Reagent, MilliporeSigma
Protein Extraction Protein Extraction Reagents
BugBuster 10X is a concentrated formulation of the proprietary detergents employed in BugBuster without the addition of salts or buffer components.
  • Bioprocessing benefits
  • Freedom to control pH
  • Maximum extraction and activitiy of your target protein
  • Allows user-defined dilution
This has all of the bioprocessing benefits of standard BugBuster in addition to the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein. Concentrated BugBuster provides a flexible alternative to the ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components.