Presto® Highly Dynamic Temperature Control Systems, JULABO
Supplier: JULABO
9420401.14 9420401.33 9421401.14 9421401.33 9420801.14 9421801.14 9420801.16.T 9421801.16.T 9420300.14
10790-450EA 73946.15 CAD
10790-450 10790-452 10790-454 10790-456 10790-458 10790-460 10790-462 10790-464 10790-514
Presto® Highly Dynamic Temperature Control Systems, JULABO
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
An entirely new generation of highly dynamic temperature control systems by JULABO. The new PRESTO® systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
  • For highly precise, external temperature applications
  • Rapid heating and cooling
  • Fast compensation of exothermic reactions
  • Wide working temperature ranges without changing fluids
  • Highest performance with small footprint
  • Space-saving design optimizes space utilization in your lab
  • 5.7” industrial color TFT touch screen
  • Well-organized view of important information with unmatched, intuitive user friendliness
  • Up to 3 user levels with password management
  • USB (Host and Device), Ethernet, SD-Card slot, RS232 / optional RS485 / optional Profibus DP
  • Stand-by input
  • Filling system accessible from the top
  • Removable venting grid for simplified removal of dust
These instruments cover a working temperature of -80 to 250 °C with high cooling and heating capacity. Highly efficient components give these instruments the ability to compensate for exothermic and endothermic reactions with extraordinary speed. Permanent internal monitoring and self-lubricating pumps contribute to the new PRESTO®’s long service life.
The integrated 5.7-inch color industrial-grade touch panel is one of the identifying characteristics of the new PRESTO®. It gives the user a clear and well-organized view of important information while greatly improving user-friendliness. Once in operation, the new PRESTO® units are whisper quiet and barely audible in a laboratory. The new PRESTO® units are extremely robust and work reliably even if the ambient temperature climbs as high as 40°C. Multiple interfaces permit remote control of the PRESTO® across networks and in superordinated control systems. Closed side panels without ventilation slits reduces operational space to a minimum.
Caution: Use only with approved JULABO Fluids.
Certifications: CE.
Accessories information: Stainless steel threaded adapters and fittings for connection to Julabo temperature control units.
Ordering information: Use only with approved JULABO Fluids; Thermal P60 (-60 to 250°C; 89421-992, 89421-994) or Thermal P90 (-90 to 170°C; 89421-996, 89421-998). Systems can operate with fluorinated fluids such as Fluorinert® and Galden® fluids. Additional accessories are available.