KOD XL DNA Polymerase, MilliporeSigma

Supplier: MilliporeSigma

71087-3 71087-4
CA80511-388EA 1502.39 CAD
CA80511-388 CA80511-390
KOD XL DNA Polymerase, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits

KOD XL DNA Polymerase is an optimized blend of KOD DNA Polymerase and a mutant form of KOD that is deficient in 3′→5′ exonuclease activity.


  • Ideal for amplification of large DNA fragments from purified DNA or crude samples
  • Amplifies DNA templates up to 30 kbp
  • Successfully amplifies GC-rich sequences
  • Efficiently incorporates derivatized dNTPs


The enzyme mixture is designed for reliable amplification of long, complex targets with robust yield and high accuracy. It can also be used for incorporation of derivatized dNTPs in PCR amplicons. It generates a mixture of PCR products with blunt and 3′-nucleotide overhangs, suitable for cloning with various vector kits.

Order Now

SPECIFICATIONS

Environmentally Preferable
Storage conditions −20 °C
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR