Low Ash Quantitative Paper Grade 391, Ahlstrom
Supplier: Ahlstrom
3910-0425 3910-0470 3910-0550 3910-1250 3910-1500 3910-1850 3910-4000 3910-0700 3910-0900 3910-1100 3910-2400 3910-2700 3910-3200
10754-440PK 44.24 CAD
10754-440 10754-442 10754-444 10754-446 10754-448 10754-450 10754-452 10754-508 10754-510 10754-512 10754-514 10754-516 10754-518
Low Ash Quantitative Paper Grade 391, Ahlstrom
Filters Filter Papers Quantitative Filter Paper
Ahlstrom Grade 391 is a fine retention low ash (Ash 0.01%) filter paper suitable for routine quantitative gravimetric techniques.
It is used for filtration of fine particles and has a slow flow rate as a result.