Perfect DNA™ Markers, MilliporeSigma
Supplier: MilliporeSigma
70540-3
69002-3
70087-3
CA80030-890EA
368.79
CAD
CA80030-890
CA80030-932
CA80031-040
Perfect DNA™ Markers, MilliporeSigma
Electrophoresis Reagents
Molecular Weight Markers
Easy-to-remember sizes for routine use in plasmid cloning.
- Easy-to-remember sizes
- Uniform band intensities
- Instant band identification
The Perfect DNA™ Markers contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities except for the easily identifiable reference bands, which are useful for instant band identification.
Delivery information: An extra vial of 6X DNA Gel Loading Buffer is included.