Anti-ETS2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA526845
CAPIPA526845EA 3406.91 CAD
CAPIPA526845
Anti-ETS2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and mouse based on sequence homology. ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ETS2 (v-ets avian erythroblastosis virus E26 oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR