Anti-SLC25A37 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA526720
CAPIPA526720EA 3335.52 CAD
CAPIPA526720
Anti-SLC25A37 Rabbit Polyclonal Antibody
Antibodies
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:25, Immunocytochemistry: 1:25, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SLC25A37 (solute carrier family 25 (mitochondrial iron transporter), member 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR