Anti-RNF40 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
MA1014D650
CAPIMA1014D650EA 2208.41 CAD
CAPIMA1014D650
Anti-RNF40 Rabbit Polyclonal Antibody
Antibodies
For Western Blot, this antibody has non-specific bands at 65 kDa. The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.

Recommended Dilutions: Western Blot: 1 µg/ml, Immunofluorescence: 1:200, Immunoprecipitation: Assay-Dependent

Type: Primary
Antigen: RNF40 (ring finger protein 40, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR