Anti-VDAC2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89782
89298-094EA 989.25 CAD
89298-094
Anti-VDAC2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to VDAC2

Approx 30kDa band observed in Human Brain and Human Testis lysates (calculated MW of 31.6kDa according to NP_003366.2). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: VDAC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR