Anti-CBX5 / HP1-Alpha (chromobox homolog 5) Goat Polyclonal Antibody

Supplier: GeneTex
GTX89432
89297-626EA 989.25 CAD
89297-626
Anti-CBX5 / HP1-Alpha (chromobox homolog 5) Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CBX5 / HP1-Alpha

Approx. 22kDa band observed in lysates of Human fibroblast cell lines A431 and HeLa (calculated MW of 22.2kDa according to NP_036249.1). Recommended concentration: 0.03-0.1µg/ml. In paraffin embedded Human Uterus shows strong nuclear staining in the glandular epithelial cells. Recommended concentration, 2-3µg/ml. Variants (NP_036249.1; NP_001120793.1; NP_001120794.1) encode the same protein.

Type: Primary
Antigen: CBX5 / HP1-Alpha (chromobox homolog 5)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR