Anti-TIP47 (perilipin 3) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85491
89329-572EA 1005.2 CAD
89329-572
Anti-TIP47 (perilipin 3) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TIP47

Type: Primary
Antigen: TIP47 (perilipin 3)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR