Anti-CCDC98 (family with sequence similarity 175, member A) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85433
89329-516EA 980.99 CAD
89329-516
Anti-CCDC98 (family with sequence similarity 175, member A) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CCDC98

Type: Primary
Antigen: CCDC98 (family with sequence similarity 175, member A)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR