Anti-KLHL2 Mouse Monoclonal Antibody [clone: 1G7]

Supplier: GeneTex
GTX84259
89273-312EA 1147.02 CAD
89273-312
Anti-KLHL2 Mouse Monoclonal Antibody [clone: 1G7]
Antibodies
Mouse monoclonal antibody [1G7] to KLHL2

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KLHL2
Clonality: Monoclonal
Clone: 1G7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR