Anti-HSP84 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX79247
89291-964EA 1003.6 CAD
89291-964
Anti-HSP84 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Heat Shock Protein 84

Recommended Starting Dilutions: ICC/IF: 10 - 20 µg/ml IHC: 1:40 - 1:500 WB: 1:500 - 1:20000 IP: 2 µg IHC-Fr: 10 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HSP84
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR