Anti-BARHL2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77863
89263-660EA 994.88 CAD
89263-660
Anti-BARHL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to BARHL2

WB: Suggested dilution at 0.05-2.0 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: BARHL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR