Anti-P53 Mouse Monoclonal Antibody [clone: FP3-2 [FPS392]]

Supplier: GeneTex
GTX75422
89328-660EA 995.65 CAD
89328-660
Anti-P53 Mouse Monoclonal Antibody [clone: FP3-2 [FPS392]]
Antibodies
Mouse monoclonal [FP3-2] to human p53 ( P392 )

For IHC-P, IP and WB: Use at an assay dependent dilution. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher. * This product requires antigen retrieval using heat treatment prior to staining of paraffin sections.

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: FP3-2 [FPS392]
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR