Anti-GLS2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX46870
89288-660EA 998.96 CAD
89288-660
Anti-GLS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GLS2 (Middle)

Recommended Dilutions: For Western Blot: Use at 2.5 ug/ml. For IHC-P: Use at 4-8 ug/mL.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: GLS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol GLS2
Antigen name Glutaminase 2 (liver mitochondrial)
Environmentally Preferable
Antigen synonyms GLS|GLS2|hLGA|L-glutaminase|LGA|mitochondrial|Glutaminase liver isoform|GA
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 27165
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide
Purification Protein A Purified Total IgG
Concentration 1 mg/ml
Storage buffer Phosphate buffered saline (PBS)
Storage temperature Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR