Anti-ATPase H+ transporting V0 Subunit e2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45821
89268-078EA 986.33 CAD
89268-078
Anti-ATPase H+ transporting V0 Subunit e2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATP6V0E2 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR