Anti-C1ORF92 (leucine rich repeat containing 71) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45711
89267-868EA 986.6 CAD
89267-868
Anti-C1ORF92 (leucine rich repeat containing 71) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C1ORF92 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: C1ORF92 (leucine rich repeat containing 71)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR