Anti-GNIP / TRIM7 (tripartite motif containing 7) Goat Polyclonal Antibody

Supplier: Genetex
GTX24541
89288-092EA 931.87 CAD
89288-092
Anti-GNIP / TRIM7 (tripartite motif containing 7) Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to GNIP / TRIM7

Approx 25-30kDa band observed in Human Skeletal Muscle lysate (predicted MW of 25kDa according to NP_203128). Recommended for use at 1µg/ml.

Type: Primary
Antigen: TRIM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR