Anti-PRDM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100093
89318-160EA 948.4 CAD
89318-160
Anti-PRDM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRDM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR