4X SDS Sample Buffer, MilliporeSigma
Supplier: MilliporeSigma
70607-3 70607-3
CA80108-396EA 106.15 CAD
CA80108-396 EM70607-3
4X SDS Sample Buffer, MilliporeSigma
Buffers Electrophoresis Buffers Loading Buffers
The 4X SDS Sample Buffer is a standard formulation commonly used for SDS-PAGE analysis of proteins
The solution includes DTT for complete denaturation of disulfide bonds. The buffer can be used at 2X for most applications.