Pellet Paint® NF Co-Precipitant, MilliporeSigma

Supplier: MilliporeSigma

70748-3 70748-4
CA80109-662EA 325.45 CAD
CA80109-662 CA80509-744
Pellet Paint® NF Co-Precipitant, MilliporeSigma
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Pellet Paint NF Co-Precipitant is a nonfluorescent dye-labeled carrier compatible with fluorescent sequencing.


  • Efficient and rapid precipitation of sequencing products
  • Efficient removal of dye terminators
  • Direct visualization and tracking of precipitated material
  • No effect on sequencing reaction
  • Substitute for original Pellet Paint® Co-Precipitant for fluorescent detection applications


It facilitates rapid removal of BigDye Terminators during the alcohol precipitation of cycle sequencing reaction products


To avoid extra sample handling, this can be added directly to the reaction mix, template DNA, crude PCR samples, or dilution buffer before the cycle sequencing reaction. Although it absorbs in the UV range, accurate spectrophotometric measurements of DNA or RNA samples are possible.


Cycle sequencing reactions can be precipitated rapidly with 1 µl of carrier per reaction and centrifugation times of 10 minutes. Resuspension of pelleted sequencing reaction products in deionized formamide can be confirmed by checking for dissolution of the carrier.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule DNA,RNA
Refine target molecule
Sample type
Method / Format Precipitation
Storage conditions -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR