Harmonic Motion Spring
Supplier: Wards
PH0713
470157-640EA 114.85 CAD
470157-640
Harmonic Motion Spring
Educational Materials Physics Educational Materials Mechanics Learning Activities
Ideal for demonstrations or hands on experiments in the physics classroom
  • Use it to investigate Simple Harmonic Motion or Hooke’s Law
  • The spring is 7 inches (18 cm) long with a maximum diameter of 1.4 inches (3.5 cm) and a spring constant (k) of 9 to 9.5 N/m
The Harmonic Motion spring’s elongations are closely proportioned to the applied forces, making it easy to use. The windings of this truncated cone, made of stainless steel wire, interfere with each other less than those of a helical spring, making the motion truly sinusoidal. The spring constantMore Product Information