Beaded Pipe to Standard Taper Joint Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5003-42 5003-44 5003-50 5003-52 5003-10 5003-12 5003-14 5003-20 5003-22 5003-24 5003-30 5003-32 5003-33 5003-40 5003-54 5003-60 5003-62 5003-63
10767-168EA 134.22 CAD
10767-168 10767-170 10767-172 10767-174 10767-714 10767-716 10767-718 10767-720 10767-722 10767-724 10767-726 10767-728 10767-730 10767-732 10767-734 10767-736 10767-738 10767-740
Beaded Pipe to Standard Taper Joint Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This transition adapter is designed to convert beaded pipe to a standard taper joint.
  • Available with inner [ST] joint or outer [ST] joint
  • Made of borosilicate glass