Premium Bosshead with Universal Swivel

Supplier: EISCO SCIENTIFIC LLC
PRBH04
470165-428EA 22.95 CAD
470165-428
Premium Bosshead with Universal Swivel
Clamps Bossheads
This bosshead can swivel 360°.

  • One-Piece Clamping Screws
  • High-strength, Powder-Coated Alloy Construction
  • All-Metal Material to Prevent Torque Damage
  • Bosshead Holds Rods up to 25 mm in Diameter

It has one-piece, threaded metal screws and an all-metal construction to carry heavy loads and prevent torque damage.

It’s made of a powder-coated alloy and can hold rods up to 25 mm in diameter.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR