Premium Three-Finger Clamp, Cork Lined
Supplier: EISCO SCIENTIFIC LLC
PRCMP09
470165-418EA 44.05 CAD
470165-418
Premium Three-Finger Clamp, Cork Lined
Clamps General Purpose Clamps
This three-finger clamp is made of powder-coated steel.
  • Maximum Clamp Opening: 8.5 cm
  • Connecting Rod: 14 cm
  • Powder-Coated Steel Construction
  • Cork-Lined
  • Thread-Fitted Actuating Joints
The clamp is cork-lined and attached to a rugged, aluminum rod.
It has thread-fitted actuating joints and a 10 mm-diameter connecting rod.