Premium Three-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
Supplier: Wards
PRCMP06 PRCMP06BH
470165-378EA 52.45 CAD
470165-378 470165-380
Premium Three-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
Clamps General Purpose Clamps
Heavy-Duty, Spring Loaded Clamp Jaw.
  • Made of steel and powder-coated alloy
  • All metal adjustment screws
  • Three vinyl coated clamp fingers
This three-prong clamp has a 6 cm maximum clamp opening and a 12.5 cm steel connecting rod.