Premium Two-Prong Clamp with Bosshead, Vinyl Coated
Supplier: EISCO SCIENTIFIC LLC
PRCMP05BH
470165-376EA 45.9 CAD
470165-376
Premium Two-Prong Clamp with Bosshead, Vinyl Coated
Clamps General Purpose Clamps
Vinyl Lined Extension Lab Clamp
  • Maximum clamp opening is 11 cm
  • Bosshead accommodates up to 19 mm diameter rod
Clamp constructed of powder coated steel, two vinyl coated prongs attached to a high strength alloy swivel bosshead for ease of use in the lab.