Premium Two-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
Supplier: Wards
PRCMP04 PRCMP04BH
470165-406EA 54.35 CAD
470165-406 470165-408
Premium Two-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
Clamps General Purpose Clamps
This durable clamp is made of powder-coated steel.
  • Maximum Clamp Opening: 7 cm
  • Connection Rod: 12.5 cm
  • Spring-Loaded Clamp Jaw
  • Two Vinyl-Coated Prongs
  • Powder-Coated Steel Construction
The clamp has thread-fitted actuating joints and two vinyl-coated prongs.
A spring-loaded clamp jaw with a dual-screw adjustment can fine-tune any fitting.
Order Now
SPECIFICATIONS
Environmentally Preferable