Premium Four-Prong Clamp, Cork Lined
Supplier: Wards
PRCMP02 PRCMP02BH
470165-398EA 43.75 CAD
470165-398 470165-400
Premium Four-Prong Clamp, Cork Lined
Clamps General Purpose Clamps
Cork Lined Extension Lab Clamp
  • Maximum clamp opening is 10.5 cm
  • 14 cm stainless steel connecting rod
Clamp constructed of powder coated steel, four cork lined prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.