Premium Three-Finger Clamp, Vinyl Coated
Supplier: EISCO SCIENTIFIC LLC
PRCMP01 PRCMP01BH
470165-374EA 40.35 CAD
470165-374 470165-396
Premium Three-Finger Clamp, Vinyl Coated
Clamps General Purpose Clamps
This premium lab clamp features thread-fitted actuating joints.
  • Maximum Clamp Opening: 8.5 cm
  • Connecting Rod: 14 cm
  • Powder-Coated Steel Construction
  • Vinyl-Coated Fingers
  • Thread-Fitted Actuating Joints
The clamp is made of powder-coated steel.
Thread-fitted actuating joints join the vinyl-coated fingers to a 10 mm connecting rod.