A-Shaped Base Retort Stand
Supplier: Wards
STDRDAB01 STDRDAB02 STDRDAB03
470165-368EA 63.5 CAD
470165-368 470165-370 470165-372
A-Shaped Base Retort Stand
Retort Stands
This cast iron, powder-coated, A-shaped base includes a stainless steel rod.
  • Powder-Coated
  • Heavy-Duty
  • A-Shaped
  • Multiple Sizes Available
  • Stainless Steel Rod Included
These bases can be nested in one another.
Multiple sizes are available.