Allylthiourea, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08158.0050 8.08158.0250 8.08158.0050 8.08158.0250
EM8.08158.0050CS 14371.68 CAD
EM8.08158.0050 EM8.08158.0250 CA8.08158.0050 CA8.08158.0250
Allylthiourea, Sigma-Aldrich®
Allylthiourea
Formula: H₂CCHCH₂NHC(S)NH₂
MDL Number: MFCD00004940
CAS Number: 109-57-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

CAS 109-57-9
Synonyms Thiosinamine
Hill Formula C₄H₈N₂S
Molar Mass 116.19 g/mol
Melting Point 70 - 73 °C
Bulk density 550 kg/m³
Solubility 66 g/l (20 °C)
LD 50 oral LD50 Rat 200 mg/kg
Assay (HPLC, area%) ≥ 98.0 % (a/a)
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR