TexWrite® 22 Cleanroom Spiral Notebooks, Texwipe®
Supplier: Texwipe
TexWrite®
TX5742 TX5740 TX5741
TWTX5740CS 676.08 CAD
TWTX5740 TWTX5741 TWTX5742
TexWrite® 22 Cleanroom Spiral Notebooks, Texwipe®
Notebooks
These spiral-bound notebooks are an economical choice for note taking and data recording in the cleanroom.
  • Printed with low sodium, IPA resistant inks
  • Lie flat and pages rotate freely for ease of use
  • High-density polyethylene covers
  • White, college-ruled with 100 writable pages (50 sheets)
Ordering information: Notebooks are cleanroom packaged using Bag-Within-A-Bag® technology, permits alcohol wipedown of exterior bag before introduction of inner bag into cleanrooms, sterile suites and aseptic environments.