Human Recombinant IGF-I (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-34-100UG 100-34-1MG 100-34-20UG 100-34-5MG
10787-530EA 354.2 CAD
10787-530 10787-532 10787-534 10821-896
Human Recombinant IGF-I (from E. coli)
Proteins and Peptides
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death. Mature human IGF-I is 100% homologous to bovine and porcine IGF-1 proteins.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at –20°C upon delivery.