Human Recombinant FGF-basic 147 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-28-10UG 100-28-1MG 100-28-50UG 100-28-500UG 100-28-100UG
10787-482EA 141.54 CAD
10787-482 10787-484 10787-486 10821-760 10821-968
Human Recombinant FGF-basic 147 (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (bFGF), also known as FGF2, is expressed by endothelial cells and is a mediator of angiogenesis. bFGF also has cardioprotective functions during heart injury. The application of bFGF is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Degredation of the full length bFGF N-terminus results in a truncated bFGF147aa protein, when the protein is isolated from biological sources. The N-terminus extensions influence the localization of bFGF within the cell, but do not affect the biological activity of bFGF. Therefore, there are no detectable differences in biological activity between the full length bFGF-154aa and the truncated bFGF-147aa recombinant proteins.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at −20 °C upon delivery.