Rat Tunnel™ Stainless Steel End Caps, Certified, Bio-Serv
Supplier: Bio-Serv
K3326-ECM K3325-ECM
CA10798-826EA 150.23 CAD
CA10798-826 CA10798-828
Rat Tunnel™ Stainless Steel End Caps, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high-temp polycarbonate with stainless steel end caps
An excellent enrichment option for toxicology studies. Transparency makes routine cage checks easier and faster. The stainless steel end caps prevent gnawing of polycarbonate. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).