Ultimate Turf Foraging Boards, Bio-Serv
Supplier: Bio-Serv
K3575 K3576 K3577
CA10798-816EA 37.21 CAD
CA10798-816 CA10798-818 CA10798-820
Ultimate Turf Foraging Boards, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high-density polyethylene (HDPE)
HDPE is known for its large strength-to-density ratio. Boards are strong enough for use in cages. Add treats or peanut butter to turf to increase foraging opportunities. Equipped with hole for hanging option. Cagewasher safe.