Vinyl GHS Labels "Pictogram"
Supplier: EMEDCO
GHMLSIINQ58 GHMLSIDAQ58 GHMLSIMUQ58 GHMLSICRQ58 GHMLSITOQ58 GHMLSICBQ58 GHMLSIENQ58 GHMLSIEXQ58 GHMLSIGZQ58
470165-344EA 41.2 CAD
470165-344 470165-346 470165-348 470165-350 470165-352 470165-354 470165-356 470165-358 470165-392
Vinyl GHS Labels "Pictogram"
Labels
These labels help identify hazardous items in your facility.
  • Labels stick to any surface
  • OSHA Standard-Compliant
  • Labels don't tear or peel
Die-cut GHS pictogram labels are pre-printed with the OSHA GHS required pictogram symbols. They have a white background with a red border and black graphics.
Ordering information: Each symbol is sold separately.