Human Recombinant CD40-Ligand (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
CD40-Ligand, or CD154, is a membrane glycoprotein and differentiation antigen that is expressed on the surface of T cells, monocytes, basophils, eosinophils, platelets, dendritic cells, and endothelial cells. CD40-Ligand binds and activates the CD40 receptor on antigen-presenting cells. CD40-Ligand stimulates B cell proliferation, immunoglobulin class switching, and antibody secretion. CD40-Ligand also induces cytokine production and tumoricidal activity in peripheral blood monocytes. CD40-Ligand is a co-stimulator of activated T cell proliferation and inflammatory protein production.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.