VWR® Low Form Griffin Beakers
Supplier: VWR International
10754-696 10754-698 10754-700 10754-760 10754-762 10754-946 10754-948 10754-950 10754-952 10754-954 10754-956 10754-958 10754-960 10754-962 75870-432
10754-696CS 507.46 CAD
10754-696 10754-698 10754-700 10754-760 10754-762 10754-946 10754-948 10754-950 10754-952 10754-954 10754-956 10754-958 10754-960 10754-962 75870-432
VWR® Low Form Griffin Beakers
Beakers
These low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass.
  • Double capacity scale
  • Uniform wall thickness
  • Very good chemical resistance
  • High temperature resistance
  • Meets ASTM E960, E960-93
They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics. They are printed with white graduated scale and a durable marking area.