VWR® Media/Storage Bottles with GL Screw Caps
Supplier: VWR International
10754-814 10754-816 10754-818 10754-820 10754-822 10754-824 10754-826 10754-828 10754-830 10754-832 10754-834 10754-836 10754-838 10754-840 75993-582 75993-584 76316-244 76316-246
10754-814CS 256.52 CAD
10754-814 10754-816 10754-818 10754-820 10754-822 10754-824 10754-826 10754-828 10754-830 10754-832 10754-834 10754-836 10754-838 10754-840 75993-582 75993-584 76316-244 76316-246
VWR® Media/Storage Bottles with GL Screw Caps
Bottles Glass Bottles
Media/storage bottles are designed with a wide opening (30 mm Int.Ø) and a drip-free polypropylene pour ring.
  • Very good chemical resistance
  • High thermal-shock resistance
  • Scale graduations
  • Autoclavable
It can be autoclaved to 180 °C (356 °F). They feature enhanced graduations and marking spots printed in permanent white color.
Certifications:
These bottles are manufactured from high quality Type 1, Class A, 33 expansion Borosilicate glass.
Ordering information: All bottles are available with or without a GL 45 polypropylene cap, with theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable