Mouse Recombinant IL-12 (from HEK293 Cells)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-25-100UG 200-25-1MG 200-25-500UG
10788-004EA 2831.79 CAD
10788-004 10788-006 10788-008 10788-010 10822-142
Mouse Recombinant IL-12 (from HEK293 Cells)
Proteins and Peptides
Interleukin 12 (IL-12) is an inducer of cell-mediated immunity and is produced by dendritic cells, macrophages, and B cells after antigen stimulation. IL-12 functions to promote naïve T cell differentiation into the type 1 T helper cell (Th1) lineage. IL-12 also stimulates the production of interferon gamma (IFN-γ) and tumor necrosis factor alpha (TNF-α) from T cells and natural killer cells. Mouse IL-12 is active on human and mouse cells. 
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.