Mouse Recombinant FOLLISTATIN (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Resistin-like molecule-alpha (RELM-α) is a member of the RELM family of secreted proteins containing conserved C-terminus cysteines. The RELM family consists of Resistin (FIZZ3), RELM-α (FIZZ1), RELM-β (FIZZ2), and RELM-γ (FIZZ4). RELM-α and Resistin are secreted by adipocytes, unlike RELM-β which is secreted by gastrointestinal epithelial cells, and RELM-γ which is expressed by peripheal blood granulocytes and bone marrow cells.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.