Mouse Recombinant GRO-A/CXCL1 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-23-100UG 200-23-1MG 200-23-20UG 200-23-5UG 200-23-500UG
10787-988EA 884.72 CAD
10787-988 10787-990 10787-992 10787-994 10822-036
Mouse Recombinant GRO-A/CXCL1 (from E. coli)
Proteins and Peptides
Growth regulated protein alpha (GRO-α), also known as CXCL1, is a chemokine that has mitogenic properties and is a neutrophil chemoattractant. GRO-α is secreted by macrophages, epithelial cells, neutrophils, and melanomas. GRO-α signals through the CXCR2 chemokine receptor and is important during spinal cord formation, inflammation, angiogenesis, tumorigenesis, and wound healing.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.