Mouse Recombinant IL-13 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Interleukin 13 (IL-13) is a cytokine secreted from type 2 T helper (Th2) cells. IL-13 has overlapping functions with interleukin 4 (IL-4), including the induction of immunoglobulin E (IgE) secretion from B cells, and the inhibition of interleukin 1 beta (IL-1β), tumor necrosis factor alpha (TNF-α), interleukin 8 (IL-8), and interleukin 6 (IL-6) inflammatory cytokine expression. IL-13 also regulates immune cell inflammation in response to the pathophysiological changes of surrounding non-immune cells. The IL-13 receptor consists of the IL-4Ra and IL-13Ra1 subunits. IL-13 can also bind the IL-13Ra2 receptor with high affinity. IL-13 functions are mediated through the JAK/STAT signaling pathway. Human and mouse IL-13 are cross-reactive.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.