HydroLogix™ ART Pipette Tips, Gel Loading, Thermo Scientific
Supplier: Thermo Scientific
3621 3631
53225-648CS 902.6 CAD
53225-648 53225-652
HydroLogix™ ART Pipette Tips, Gel Loading, Thermo Scientific
Pipette Tips
Designed for loading vertical electrophoresis gels.
  • Choice of round or flat tip
  • Intended for loading gels of three thicknesses to suit specific applications
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)