Mouse Recombinant IL-4 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-18-100UG 200-18-20UG 200-18-1MG 200-18-5UG 200-18-500UG
10789-136EA 920.92 CAD
10789-136 10789-138 10789-140 10789-142 10822-100
Mouse Recombinant IL-4 (from E. coli)
Proteins and Peptides
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4Rα receptor to activate STAT6 signaling.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.