Mouse Recombinant IL-2 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-17-100UG 200-17-1MG 200-17-20UG 200-17-5UG 200-17-500UG
10788-764EA 891.88 CAD
10788-764 10788-766 10788-768 10788-770 10822-086
Mouse Recombinant IL-2 (from E. coli)
Proteins and Peptides
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.